Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Commd8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106004.2 | NP_001099474.1 | 2 - 3 | 234 | 116 | |
GenBank mRNA | BC159414.1 | - | 2 - 3 | 234 | 8368074 | 116 |
FQ212906.1 | - | 2 - 3 | 243 | 116 | ||
FQ221260.1 | - | 2 - 3 | 235 | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |