Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nhsl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002728626.4 | XP_002728672.1 | 1 - 2 | 1701 | 103 | |
XM_006227668.2 | XP_006227730.1 | 1 - 2 | 1701 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |