Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ankdd1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006243300.2 | XP_006243362.1 | 14 - 15 | 2761 | 54 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Rattus norvegicus 1700 Probe spans exons |