Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rtn4ip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107644.3 | NP_001101114.1 | 3 - 4 | 889 | 90 | |
XM_006256589.2 | XP_006256651.1 | 3 - 4 | 910 | 90 | ||
XM_006256590.2 | XP_006256652.1 | 3 - 4 | 910 | 90 | ||
XM_008772999.1 | XP_008771221.1 | 4 - 5 | 578 | 90 | ||
GenBank mRNA | BC082054.1 | - | 3 - 4 | 910 | 90 | |
BC166465.1 | - | 3 - 4 | 893 | 9030981 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus Probe spans exons |