Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnajc28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014124.1 | NP_001014146.1 | 1 - 2 | 213 | 103 | |
GenBank mRNA | BC079055.1 | - | 1 - 2 | 213 | 7115421 | 103 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Rattus norvegicus Inventoried Probe spans exons |