Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gabpa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108841.1 | NP_001102311.1 | 1176 | 88 | ||
XM_008768584.1 | XP_008766806.1 | 5 - 6 | 785 | 88 | ||
XM_008768585.1 | XP_008766807.1 | 5 - 6 | 780 | 88 | ||
XM_008768586.1 | XP_008766808.1 | 5 - 6 | 780 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |