Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Galntl6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001135756.1 | NP_001129228.1 | 5 - 6 | 896 | 86 | |
XM_008771227.1 | XP_008769449.1 | 4 - 5 | 1550 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |