Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vps28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001130492.1 | NP_001123964.1 | 4 - 5 | 263 | 77 | |
GenBank mRNA | BC168742.1 | - | 4 - 5 | 209 | 6919204 | 77 |
FQ225519.1 | - | 4 - 5 | 230 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |