Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Znf7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006241919.2 | XP_006241981.1 | 3 - 4 | 436 | 64 |
Gene Symbol: Zfp7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001142757.1 | NP_001136229.1 | 3 - 4 | 282 | 64 | |
XM_006241928.2 | XP_006241990.1 | 3 - 4 | 436 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Rattus norvegicus Probe spans exons |