Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Maf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014085.1 | NP_001014107.1 | 4 - 5 | 731 | 79 | |
XM_006241811.1 | XP_006241873.1 | 4 - 5 | 806 | 79 | ||
XM_006241812.1 | XP_006241874.1 | 4 - 5 | 775 | 79 | ||
GenBank mRNA | BC083712.1 | - | 4 - 5 | 731 | 7190634 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus Probe spans exons |