Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ybey | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108529.1 | NP_001101999.1 | 2 - 3 | 316 | 127 | |
XM_006256301.2 | XP_006256363.1 | 2 - 3 | 327 | 127 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |