Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spdef | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109530.1 | NP_001103000.1 | 1 - 2 | 342 | 89 | |
XM_006256229.2 | XP_006256291.1 | 1 - 2 | 1102 | 89 | ||
GenBank mRNA | BC166744.1 | - | 1 - 2 | 346 | 7938752 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus Probe spans exons |