Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spdef | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109530.1 | NP_001103000.1 | 2 - 3 | 771 | 69 | |
XM_006256229.2 | XP_006256291.1 | 2 - 3 | 1531 | 69 | ||
XM_008772827.1 | XP_008771049.1 | 2 - 3 | 641 | 69 | ||
GenBank mRNA | BC166744.1 | - | 2 - 3 | 775 | 7938752 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Rattus norvegicus Probe spans exons |