Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trmt5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108713.1 | NP_001102183.1 | 3 - 4 | 764 | 71 | |
XM_006240213.2 | XP_006240275.1 | 2 - 3 | 740 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Rattus norvegicus Probe spans exons |