Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mpv17l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001098240.1 | NP_001091710.1 | 1 - 2 | 320 | 78 | |
GenBank mRNA | BC091193.1 | - | 1 - 2 | 320 | 7373705 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |