Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Six4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106739.1 | NP_001100209.1 | 3 - 4 | 684 | 82 | |
XM_006240207.2 | XP_006240269.1 | 1 - 2 | 848 | 82 | ||
XM_006240208.2 | XP_006240270.1 | 2 - 3 | 1048 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |