Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snapc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108033.1 | NP_001101503.1 | 5 - 6 | 757 | 73 | |
GenBank mRNA | BC161969.1 | - | 5 - 6 | 776 | 9028949 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |