Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sypl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014263.1 | NP_001014285.1 | 3 - 4 | 304 | 81 | |
XM_006240001.2 | XP_006240063.1 | 3 - 4 | 317 | 81 | ||
XM_006240002.2 | XP_006240064.1 | 3 - 4 | 317 | 81 | ||
GenBank mRNA | BC082078.1 | - | 3 - 4 | 304 | 7133263 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |