Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snapin | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001170576.1 | NP_001164047.1 | 1 - 2 | 171 | 65 | |
GenBank mRNA | DY315040.1 | - | 1 - 2 | 176 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Rattus norvegicus Probe spans exons |