Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cbx4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008768485.1 | XP_008766707.1 | 5 | 1635 | 93 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Rattus norvegicus 1700 Both primers and probe map within a single exon |