Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RT1-M5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001048045.2 | NP_001041510.2 | 4 - 5 | 896 | 132 | |
NM_001168353.1 | NP_001161825.1 | 4 - 5 | 896 | 132 | ||
GenBank mRNA | EF638822.1 | - | 3 - 4 | 804 | 132 | |
EF638823.1 | - | 3 - 4 | 804 | 132 | ||
EF638824.1 | - | 3 - 4 | 804 | 132 | ||
EF638825.1 | - | 3 - 4 | 804 | 132 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons |