Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdh26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191745.1 | NP_001178674.1 | 2 - 3 | 153 | 67 | |
XM_008762509.1 | XP_008760731.1 | 2 - 3 | 407 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Rattus norvegicus Probe spans exons |