Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ankrd31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008760712.1 | XP_008758934.1 | 11 - 12 | 1786 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |