Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ankrd31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008760712.1 | XP_008758934.1 | 12 - 13 | 1946 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |