Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kng1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012696.2 | NP_036828.2 | 1282 | 157 | ||
GenBank mRNA | BC087028.1 | - | 1282 | 7130388 | 157 | |
FQ211035.1 | - | 690 | 157 | |||
FQ211199.1 | - | 688 | 157 | |||
FQ219050.1 | - | 688 | 157 | |||
FQ219322.1 | - | 1164 | 157 | |||
FQ219531.1 | - | 690 | 157 | |||
M11883.1 | - | 1052 | 157 | |||
X02299.1 | - | 1017 | 157 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus Rat Inflammation Inventoried Rat_Inflammation Probe spans exons |