Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Akr1c13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014240.3 | NP_001014262.3 | 3 - 4 | 391 | 159 | |
GenBank mRNA | AB221346.1 | - | 3 - 4 | 362 | 159 | |
AY327508.1 | - | 3 - 4 | 427 | 159 | ||
BC088169.1 | - | 3 - 4 | 391 | 7322153 | 159 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |