Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zscan20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008764175.1 | XP_008762397.1 | 8 | 3394 | 141 | |
XM_008764176.1 | XP_008762398.1 | 8 | 3512 | 141 | ||
XM_008764177.1 | XP_008762399.1 | 9 | 4488 | 141 | ||
XM_008764178.1 | XP_008762400.1 | 8 | 3343 | 141 | ||
XM_008764179.1 | XP_008762401.1 | 8 | 4314 | 141 | ||
XM_575911.5 | XP_575911.4 | 9 | 4071 | 141 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Rattus norvegicus 1700 Both primers and probe map within a single exon |