Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cox7a2l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106704.1 | NP_001100174.1 | 3 | 1201 | 74 | |
XM_006239680.1 | XP_006239742.1 | 3 | 1030 | 74 | ||
GenBank mRNA | BC166896.1 | - | 3 | 1013 | 6921504 | 74 |
FQ213890.1 | - | 3 | 1013 | 74 | ||
FQ215773.1 | - | 3 | 982 | 74 | ||
FQ220707.1 | - | 3 | 1013 | 74 | ||
FQ230066.1 | - | 3 | 1013 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Transporter 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Both primers and probe map within a single exon |