Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcne1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012973.1 | NP_037105.1 | 2 | 424 | 140 | |
XM_006248034.2 | XP_006248096.1 | 2 | 437 | 140 | ||
XM_006248035.2 | XP_006248097.1 | 2 | 591 | 140 | ||
XM_008768566.1 | XP_008766788.1 | 2 | 808 | 140 | ||
GenBank mRNA | M22412.1 | - | 2 | 424 | 140 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Ion Channel 3' Most Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Both primers and probe map within a single exon |