Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arrdc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006229367.2 | XP_006229429.1 | 8 | 3760 | 153 | |
GenBank mRNA | FQ222682.1 | - | 8 | 3637 | 153 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Rattus norvegicus 1700 Both primers and probe map within a single exon |