Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mrgprh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002281.1 | NP_001002281.1 | 1 | 921 | 86 | |
XM_006227882.2 | XP_006227944.1 | 2 | 1173 | 86 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Rattus norvegicus Inventoried Both primers and probe map within a single exon |