Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Igfals | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053329.2 | NP_445781.2 | 2 | 1856 | 91 | |
GenBank mRNA | S46785.1 | - | 2 | 1856 | 91 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Both primers and probe map within a single exon |