Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bcs1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007666.1 | NP_001007667.1 | 7 - 8 | 1311 | 95 | |
GenBank mRNA | BC083660.1 | - | 7 - 8 | 1311 | 7135331 | 95 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons |