Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Krt36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008759.1 | NP_001008759.1 | 5 - 6 | 975 | 180 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Detect Genomic DNA Rattus norvegicus Probe spans exons |