Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Serpina11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008776.2 | NP_001008776.1 | 3 - 4 | 1037 | 122 | |
XM_006240481.2 | XP_006240543.1 | 4 - 5 | 1142 | 122 | ||
GenBank mRNA | AY325135.1 | - | 3 - 4 | 1077 | 122 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Detect Genomic DNA Rattus norvegicus Probe spans exons |