Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Srsf9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009255.1 | NP_001009255.1 | 1 - 2 | 252 | 148 | |
GenBank mRNA | BC087684.1 | - | 1 - 2 | 252 | 7309111 | 148 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |