Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Srsf9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009255.1 | NP_001009255.1 | 2 - 3 | 415 | 148 | |
GenBank mRNA | BC087684.1 | - | 2 - 3 | 415 | 7309111 | 148 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |