Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Enoph1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009391.1 | NP_001009391.1 | 4 - 5 | 602 | 141 | |
XM_008770023.1 | XP_008768245.1 | 4 - 5 | 652 | 141 | ||
GenBank mRNA | BC087697.1 | - | 4 - 5 | 602 | 7312419 | 141 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |