Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdh19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009448.1 | NP_001009448.1 | 4 - 5 | 780 | 155 | |
XM_006249653.2 | XP_006249715.1 | 5 - 6 | 1418 | 155 | ||
GenBank mRNA | AB121032.1 | - | 4 - 5 | 780 | 155 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |