Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cirh1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009640.1 | NP_001009640.1 | 14 - 15 | 1682 | 147 | |
XM_006255562.1 | XP_006255624.1 | 14 - 15 | 1728 | 147 | ||
XM_006255563.2 | XP_006255625.1 | 14 - 15 | 1891 | 147 | ||
GenBank mRNA | BC088461.1 | - | 14 - 15 | 1682 | 7127580 | 147 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |