Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Drg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009685.1 | NP_001009685.1 | 3 - 4 | 387 | 150 | |
GenBank mRNA | BC088410.1 | - | 3 - 4 | 387 | 7189810 | 150 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |