Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vom1r54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008963.1 | NP_001008963.1 | 852 | 106 | ||
GenBank mRNA | AY510374.1 | - | 852 | 106 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Recommended Assay Rattus norvegicus Inventoried |