Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tGap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007635.2 | NP_001007636.2 | 17 - 18 | 2414 | 149 | |
XM_006232134.2 | XP_006232196.1 | 19 - 20 | 2756 | 149 | ||
GenBank mRNA | AY631396.1 | - | 17 - 18 | 2414 | 149 | |
AY631398.1 | - | 10 - 11 | 1558 | 149 | ||
AY631399.1 | - | 17 - 18 | 2179 | 149 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |