Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fkbpl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002818.2 | NP_001002818.1 | 2 | 1173 | 95 | |
GenBank mRNA | BC086532.1 | - | 2 | 1173 | 7305083 | 95 |
FQ231484.1 | - | 2 | 1187 | 95 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Both primers and probe map within a single exon |