Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Actl9b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013893.1 | NP_001013915.1 | 1 | 1292 | 66 | |
GenBank mRNA | BC079230.1 | - | 1 | 1292 | 7132497 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus Both primers and probe map within a single exon |