Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pou3f3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_138837.1 | NP_620192.1 | 1 | 542 | 60 | |
GenBank mRNA | AJ001641.1 | - | 1 | 542 | 60 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Rattus norvegicus Inventoried Both primers and probe map within a single exon |