Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Senp8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012355.1 | NP_001012355.1 | 3 | 647 | 94 | |
XM_006243196.2 | XP_006243258.1 | 2 | 741 | 94 | ||
GenBank mRNA | BC089939.1 | - | 3 | 647 | 7310036 | 94 |
FQ226292.1 | - | 2 | 629 | 94 | ||
FQ227853.1 | - | 3 | 660 | 94 | ||
FQ231841.1 | - | 2 | 639 | 94 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Both primers and probe map within a single exon |