Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Defa9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025763.1 | NP_001020934.1 | 1 - 2 | 174 | 75 | |
GenBank mRNA | AY623753.1 | - | 1 - 2 | 174 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Rattus norvegicus Inventoried Probe spans exons |