Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcsk9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199253.2 | NP_954862.2 | 9 - 10 | 1689 | 148 | |
GenBank mRNA | AY847775.1 | - | 9 - 10 | 1689 | 148 | |
BC133063.1 | - | 9 - 10 | 1620 | 8371686 | 148 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protease Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |