Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arid5a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034934.1 | NP_001030106.1 | 4 - 5 | 373 | 100 | |
XM_006244806.2 | XP_006244868.1 | 6 - 7 | 675 | 100 | ||
XM_006244807.2 | XP_006244869.1 | 6 - 7 | 672 | 100 | ||
GenBank mRNA | BC105777.1 | - | 4 - 5 | 373 | 7931985 | 100 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Inventoried Probe spans exons |